siRNA Details
Gene Symbol: TVP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079512.3 | NP_001072980.1 | 8 | 3187 | ||
NM_001318873.1 | NP_001305802.1 | 8 | 3638 | |||
XM_011522654.2 | XP_011520956.1 | 8 | 4110 | |||
XM_017023649.1 | XP_016879138.1 | 8 | 3672 | |||
XM_017023650.1 | XP_016879139.1 | 8 | 3872 | |||
XM_017023651.1 | XP_016879140.1 | 8 | 3300 | |||
GenBank mRNA | AK124106.1 | - | 8 | 3494 | ||
AL833564.1 | - | 6 | 3073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
