siRNA Details
Gene Symbol: TVP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079512.3 | NP_001072980.1 | 6 | 807 | ||
NM_001318873.1 | NP_001305802.1 | 6 | 1258 | |||
NR_134901.1 | 7 | 877 | ||||
NR_134902.1 | 6 | 807 | ||||
NR_134903.1 | 6 | 769 | ||||
NR_134904.1 | 6 | 732 | ||||
NR_134905.1 | 5 | 662 | ||||
NR_134906.1 | 5 | 662 | ||||
XM_006720944.3 | XP_006721007.1 | 6 | 915 | |||
XM_011522654.2 | XP_011520956.1 | 6 | 915 | |||
XM_017023649.1 | XP_016879138.1 | 6 | 915 | |||
XM_017023650.1 | XP_016879139.1 | 6 | 1115 | |||
XM_017023651.1 | XP_016879140.1 | 6 | 543 | |||
GenBank mRNA | AK124106.1 | - | 6 | 1115 | ||
AK294418.1 | - | 6 | 798 | |||
AL833564.1 | - | 4 | 318 | |||
BC050569.1 | - | 6 | 582 | |||
BC146882.1 | - | 6 | 613 | 9056810 | ||
BC146886.1 | - | 6 | 613 | 9056814 | ||
BC171807.1 | - | 5 | 468 | |||
BC171816.1 | - | 5 | 468 | |||
BC171817.1 | - | 6 | 538 | 9054471 | ||
BC171819.1 | - | 7 | 683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
