siRNA Details
Gene Symbol: ZCCHC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143978.2 | NP_001137450.1 | 3 | 1912 | ||
XM_011531012.2 | XP_011529314.1 | 3 | 2049 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
