siRNA Details
Gene Symbol: ZCCHC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143978.2 | NP_001137450.1 | 3 | 2366 | ||
NR_026694.2 | 4 | 688 | ||||
XM_011531012.2 | XP_011529314.1 | 3 | 2503 | |||
GenBank mRNA | CN272796.1 | - | 4 | 616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
