siRNA Details
Gene Symbol: ZNF667-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036521.1 | 2 | 573 | |||
NR_036522.1 | 2 | 218 | ||||
GenBank mRNA | BC008651.1 | - | 2 | 159 | ||
BC110411.1 | - | 2 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
