siRNA Details
Gene Symbol: RBPMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194272.1 | NP_919248.1 | 8 | 1785 | ||
GenBank mRNA | AK124123.1 | - | 8 | 1533 | ||
AK127873.1 | - | 8 | 1801 | |||
AY369207.1 | - | 8 | 1785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
