siRNA Details
Gene Symbol: NEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130963.1 | NP_001124435.1 | 9 | 2771 | ||
NM_015257.2 | NP_056072.2 | 8 | 2552 | |||
XM_011538056.2 | XP_011536358.1 | 9 | 3451 | |||
XM_011538057.2 | XP_011536359.1 | 10 | 2873 | |||
XM_011538058.2 | XP_011536360.1 | 8 | 3229 | |||
XM_011538059.1 | XP_011536361.1 | 9 | 2625 | |||
XM_017019077.1 | XP_016874566.1 | 9 | 3222 | |||
XM_017019078.1 | XP_016874567.1 | 9 | 2485 | |||
XM_017019079.1 | XP_016874568.1 | 8 | 2675 | |||
GenBank mRNA | AB006624.2 | - | 9 | 2732 | ||
AL833310.1 | - | 9 | 2749 | |||
AL833311.1 | - | 9 | 2749 | |||
BC071597.1 | - | 9 | 2743 | |||
CR749406.1 | - | 8 | 2524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
