siRNA Details
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 23 | 4680 | ||
XM_011521831.2 | XP_011520133.1 | 23 | 4693 | |||
XM_011521832.2 | XP_011520134.1 | 23 | 4690 | |||
XM_011521833.2 | XP_011520135.1 | 22 | 4660 | |||
XM_011521834.2 | XP_011520136.1 | 22 | 4630 | |||
XM_011521835.2 | XP_011520137.1 | 21 | 4552 | |||
XM_011521836.2 | XP_011520138.1 | 23 | 4995 | |||
XM_011521837.2 | XP_011520139.1 | 21 | 4462 | |||
XM_011521839.2 | XP_011520141.1 | 10 | 3462 | |||
XM_017022439.1 | XP_016877928.1 | 22 | 4615 | |||
XM_017022440.1 | XP_016877929.1 | 22 | 4592 | |||
XM_017022441.1 | XP_016877930.1 | 21 | 4911 | |||
XM_017022442.1 | XP_016877931.1 | 16 | 4019 | |||
XM_017022443.1 | XP_016877932.1 | 15 | 4003 | |||
GenBank mRNA | AK025589.1 | - | 1 | 1533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
