siRNA Details
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 8 | 1869 | ||
NM_033544.2 | NP_291022.2 | 9 | 1953 | |||
GenBank mRNA | AK092661.1 | - | 1 | 1384 | ||
AL110104.1 | - | 1 | 581 | |||
AL527085.3 | - | 1 | 228 | |||
BC140708.1 | - | 8 | 1867 | 9021576 | ||
BC140709.1 | - | 8 | 1867 | 9021577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
