siRNA Details
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 8 | 1464 | ||
NM_033544.2 | NP_291022.2 | 9 | 1548 | |||
GenBank mRNA | AK092661.1 | - | 1 | 979 | ||
AL110104.1 | - | 1 | 175 | |||
AL110105.1 | - | 1 | 480 | |||
BC064606.1 | - | 6 | 1475 | |||
BC094739.1 | - | 6 | 2840 | |||
BC140708.1 | - | 8 | 1462 | 9021576 | ||
BC140709.1 | - | 8 | 1462 | 9021577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
