siRNA Details
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 8 | 1298 | ||
NM_033544.2 | NP_291022.2 | 9 | 1382 | |||
GenBank mRNA | AK092661.1 | - | 1 | 813 | ||
AL110105.1 | - | 1 | 314 | |||
BC064606.1 | - | 6 | 1309 | |||
BC094739.1 | - | 6 | 2674 | |||
BC113826.1 | - | 6 | 706 | |||
BC140708.1 | - | 8 | 1296 | 9021576 | ||
BC140709.1 | - | 8 | 1296 | 9021577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
