siRNA Details
Gene Symbol: MEI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152513.3 | NP_689726.3 | 6 | 644 | ||
XM_011529935.1 | XP_011528237.1 | 6 | 653 | |||
XM_011529936.1 | XP_011528238.1 | 6 | 653 | |||
XM_011529937.1 | XP_011528239.1 | 6 | 653 | |||
XM_011529938.1 | XP_011528240.1 | 6 | 653 | |||
XM_011529939.1 | XP_011528241.1 | 6 | 653 | |||
XM_011529940.1 | XP_011528242.1 | 6 | 653 | |||
XM_011529941.1 | XP_011528243.1 | 6 | 653 | |||
XM_011529942.2 | XP_011528244.1 | 6 | 844 | |||
XM_011529943.1 | XP_011528245.1 | 6 | 653 | |||
XM_011529944.1 | XP_011528246.1 | 6 | 653 | |||
XM_011529945.2 | XP_011528247.1 | 6 | 655 | |||
XM_011529946.1 | XP_011528248.1 | 6 | 653 | |||
XM_011529947.1 | XP_011528249.1 | 6 | 653 | |||
XM_011529950.1 | XP_011528252.1 | 6 | 653 | |||
XM_011529952.1 | XP_011528254.1 | 1 | 92 | |||
XM_011529955.1 | XP_011528257.1 | 6 | 653 | |||
XM_011529956.2 | XP_011528258.1 | 6 | 653 | |||
XM_017028633.1 | XP_016884122.1 | 6 | 653 | |||
GenBank mRNA | BC025400.1 | - | 6 | 660 | ||
BC035720.1 | - | 6 | 643 | |||
BC070111.1 | - | 1 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
