siRNA Details
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 3 | 438 | ||
NM_001146186.1 | NP_001139658.1 | 1 | 471 | |||
NM_006210.2 | NP_006201.1 | 4 | 514 | |||
GenBank mRNA | AF208967.1 | - | 4 | 484 | ||
AF208968.1 | - | 3 | 408 | |||
AF208970.1 | - | 1 | 472 | |||
BC052616.1 | - | 1 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
