siRNA Details
Gene Symbol: C17orf98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080465.2 | NP_001073934.1 | 3 | 355 | ||
GenBank mRNA | DY654789.1 | - | 3 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
