siRNA Details
Gene Symbol: MIDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177401.4 | NP_796375.3 | 8 | 3332 | ||
XM_005259671.3 | XP_005259728.1 | 9 | 3461 | |||
XM_005259672.3 | XP_005259729.1 | 9 | 3459 | |||
GenBank mRNA | BC015089.2 | - | 2 | 1943 | ||
BC094778.1 | - | 7 | 3229 | 30342357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
