siRNA Details
Gene Symbol: PLIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080400.1 | NP_001073869.1 | 3 | 2137 | ||
XM_006722866.2 | XP_006722929.1 | 5 | 4098 | |||
XM_006722868.3 | XP_006722931.1 | 5 | 4094 | |||
XM_011528233.2 | XP_011526535.1 | 6 | 4280 | |||
XM_017027192.1 | XP_016882681.1 | 6 | 4283 | |||
XM_017027193.1 | XP_016882682.1 | 6 | 4230 | |||
XM_017027194.1 | XP_016882683.1 | 6 | 4265 | |||
GenBank mRNA | AB067468.1 | - | 1913 | |||
BC153818.1 | - | 1 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
