siRNA Details
Gene Symbol: PLIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080400.1 | NP_001073869.1 | 3 | 1353 | ||
XM_006722866.2 | XP_006722929.1 | 5 | 3314 | |||
XM_006722868.3 | XP_006722931.1 | 5 | 3310 | |||
XM_011528233.2 | XP_011526535.1 | 6 | 3496 | |||
XM_017027192.1 | XP_016882681.1 | 6 | 3499 | |||
XM_017027193.1 | XP_016882682.1 | 6 | 3446 | |||
XM_017027194.1 | XP_016882683.1 | 6 | 3481 | |||
GenBank mRNA | AB067468.1 | - | 1129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
