siRNA Details
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017966.4 | NP_060436.4 | 5 | 1728 | ||
XM_005274077.3 | XP_005274134.1 | 5 | 1769 | |||
XM_017017955.1 | XP_016873444.1 | 2 | 1438 | |||
GenBank mRNA | AK000854.1 | - | 1 | 316 | ||
AL834261.1 | - | 5 | 1736 | |||
BC005805.2 | - | 1 | 355 | |||
BC062604.1 | - | 1 | 451 | |||
BC141827.1 | - | 5 | 1691 | 40148285 | ||
BC142702.1 | - | 5 | 1691 | 40148798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
