siRNA Details
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017966.4 | NP_060436.4 | 5 | 1425 | ||
XM_005274077.3 | XP_005274134.1 | 5 | 1466 | |||
XM_017017955.1 | XP_016873444.1 | 2 | 1135 | |||
GenBank mRNA | AK000854.1 | - | 1 | 13 | ||
AL834261.1 | - | 5 | 1433 | |||
BC005805.2 | - | 1 | 52 | |||
BC062604.1 | - | 1 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
