siRNA Details
Gene Symbol: TRANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014831.2 | NP_055646.2 | 13 | 4805 | ||
XM_006713432.3 | XP_006713495.1 | 14 | 4973 | |||
XM_011534289.2 | XP_011532591.1 | 14 | 5734 | |||
XM_017007570.1 | XP_016863059.1 | 13 | 4605 | |||
XM_017007571.1 | XP_016863060.1 | 14 | 4866 | |||
XM_017007572.1 | XP_016863061.1 | 14 | 4768 | |||
XM_017007573.1 | XP_016863062.1 | 14 | 4776 | |||
GenBank mRNA | AB002340.3 | - | 3 | 3183 | ||
AK096678.1 | - | 2 | 2421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
