siRNA Details
Gene Symbol: ULK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322500.1 | NP_001309429.1 | 29 | 3399 | ||
NM_001322501.1 | NP_001309430.1 | 28 | 3323 | |||
NM_017886.3 | NP_060356.2 | 29 | 3399 | |||
NR_136342.1 | 27 | 3234 | ||||
XM_011533874.2 | XP_011532176.1 | 29 | 3977 | |||
XM_011533880.2 | XP_011532182.1 | 13 | 1994 | |||
XM_017006711.1 | XP_016862200.1 | 28 | 3893 | |||
XM_017006712.1 | XP_016862201.1 | 29 | 3974 | |||
GenBank mRNA | AK093396.1 | - | 8 | 651 | ||
AK304341.1 | - | 29 | 3030 | |||
AL133104.1 | - | 1 | 71 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
