siRNA Details
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037225.2 | NP_001032302.1 | 3 | 338 | ||
NM_001300803.1 | NP_001287732.1 | 3 | 338 | |||
NM_001318808.1 | NP_001305737.1 | 3 | 338 | |||
XM_005273918.2 | XP_005273975.1 | 3 | 332 | |||
XM_006718502.2 | XP_006718565.1 | 3 | 332 | |||
XM_006718503.3 | XP_006718566.1 | 3 | 218 | |||
XM_006718504.2 | XP_006718567.1 | 3 | 332 | |||
XM_006718505.2 | XP_006718568.1 | 3 | 332 | |||
XM_011544940.2 | XP_011543242.1 | 3 | 213 | |||
GenBank mRNA | AK093130.1 | - | 3 | 332 | ||
BC033501.1 | - | 3 | 270 | |||
BC106951.2 | - | 2 | 133 | 40035116 | ||
BC106952.1 | - | 2 | 133 | 40035117 | ||
DB445650.2 | - | 3 | 339 | |||
HY050994.1 | - | 3 | 331 | |||
LC068587.1 | - | 1 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
