siRNA Details
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022068.3 | NP_071351.2 | 9, 10 | 1380 | ||
XM_011525723.2 | XP_011524025.1 | 9, 10 | 2178 | |||
XM_011525724.2 | XP_011524026.1 | 9, 10 | 2178 | |||
XM_011525725.2 | XP_011524027.1 | 9, 10 | 2178 | |||
XM_011525726.2 | XP_011524028.1 | 9, 10 | 2178 | |||
GenBank mRNA | AK092226.1 | - | 6, 7 | 1090 | ||
JN790819.1 | - | 9, 10 | 1214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
