siRNA Details
Gene Symbol: SPATA24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194296.1 | NP_919272.1 | 1 | 107 | ||
XM_005271916.4 | XP_005271973.1 | 1 | 224 | |||
XM_011543252.2 | XP_011541554.1 | 1 | 53 | |||
XM_011543253.2 | XP_011541555.1 | 1 | 52 | |||
XM_011543254.2 | XP_011541556.1 | 1 | 157 | |||
GenBank mRNA | AK098740.1 | - | 1 | 108 | ||
BC040037.1 | - | 1 | 87 | |||
BC050563.1 | - | 1 | 87 | 5751687 | ||
BM785501.1 | - | 1 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
