siRNA Details
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 7 | 770 | ||
XM_011544605.2 | XP_011542907.1 | 7 | 655 | |||
XM_011544606.2 | XP_011542908.1 | 7 | 706 | |||
XM_011544607.2 | XP_011542909.1 | 7 | 668 | |||
XM_017013719.1 | XP_016869208.1 | 6 | 491 | |||
XM_017013720.1 | XP_016869209.1 | 6 | 606 | |||
GenBank mRNA | AF286164.1 | - | 7 | 695 | ||
AK098787.1 | - | 7 | 771 | |||
AK291842.1 | - | 7 | 714 | |||
AK296068.1 | - | 6 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
