siRNA Details
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 9 | 953 | ||
XM_011544605.2 | XP_011542907.1 | 9 | 838 | |||
XM_011544606.2 | XP_011542908.1 | 9 | 889 | |||
XM_011544607.2 | XP_011542909.1 | 9 | 851 | |||
XM_017013718.1 | XP_016869207.1 | 8 | 704 | |||
XM_017013719.1 | XP_016869208.1 | 8 | 674 | |||
XM_017013720.1 | XP_016869209.1 | 8 | 789 | |||
GenBank mRNA | AF286164.1 | - | 9 | 878 | ||
AK291842.1 | - | 9 | 897 | |||
AK296068.1 | - | 8 | 746 | |||
AK309020.1 | - | 2 | 70 | |||
DC332022.1 | - | 2 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
