siRNA Details
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 2 | 257 | ||
XM_011544605.2 | XP_011542907.1 | 2 | 142 | |||
XM_011544606.2 | XP_011542908.1 | 2 | 193 | |||
XM_011544607.2 | XP_011542909.1 | 2 | 155 | |||
XM_017013718.1 | XP_016869207.1 | 2 | 155 | |||
XM_017013720.1 | XP_016869209.1 | 2 | 142 | |||
GenBank mRNA | AF286164.1 | - | 2 | 182 | ||
AK098787.1 | - | 2 | 258 | |||
AK291842.1 | - | 2 | 201 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031257, BC010215, AF286161, AK078212, AF418551, and others.
Gene Ontology Categories:
