siRNA Details
Gene Symbol: COA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012985.2 | NP_001013003.1 | 3 | 509 | ||
NM_001206641.2 | NP_001193570.2 | 3 | 593 | |||
NM_001301733.1 | NP_001288662.1 | 2 | 880 | |||
GenBank mRNA | BC025793.1 | - | 3 | 416 | ||
BC035925.1 | - | 3 | 506 | |||
BC116455.1 | - | 3 | 509 | 4540930 | ||
BG195363.1 | - | 1 | 196 | |||
BG204581.1 | - | 2 | 352 | |||
CB136291.1 | - | 3 | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
