siRNA Details
Gene Symbol: COA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012985.2 | NP_001013003.1 | 2 | 195 | ||
NM_001206641.2 | NP_001193570.2 | 2 | 279 | |||
NM_001301733.1 | NP_001288662.1 | 1 | 566 | |||
GenBank mRNA | BC025793.1 | - | 2 | 102 | ||
BC035925.1 | - | 2 | 192 | |||
BC116455.1 | - | 2 | 195 | 4540930 | ||
CB136291.1 | - | 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
