siRNA Details
Gene Symbol: TTC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080441.1 | NP_001073910.1 | 1 | 74 | ||
GenBank mRNA | BC146916.1 | - | 1 | 97 | 9056844 | |
BC146921.1 | - | 1 | 97 | 9056849 | ||
BC171906.1 | - | 1 | 97 | 9054560 | ||
BC171913.1 | - | 1 | 97 | 9054567 | ||
EU489483.1 | - | 1 | 136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
