siRNA Details
Gene Symbol: FRYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015030.1 | NP_055845.1 | 5 | 738 | ||
XM_005248090.4 | XP_005248147.1 | 5 | 751 | |||
XM_011513680.2 | XP_011511982.1 | 2 | 213 | |||
XM_011513681.2 | XP_011511983.1 | 2 | 213 | |||
XM_011513682.2 | XP_011511984.1 | 2 | 213 | |||
XM_011513683.2 | XP_011511985.1 | 4 | 587 | |||
XM_011513685.2 | XP_011511987.1 | 2 | 213 | |||
XM_017008046.1 | XP_016863535.1 | 6 | 1063 | |||
GenBank mRNA | AK131250.1 | - | 3 | 413 | ||
AL833444.1 | - | 5 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
