siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 76 | 13193 | ||
XM_006721206.2 | XP_006721269.1 | 76 | 13087 | |||
XM_011523146.2 | XP_011521448.1 | 77 | 13222 | |||
XM_011523147.1 | XP_011521449.1 | 77 | 13189 | |||
XM_011523148.1 | XP_011521450.1 | 77 | 13262 | |||
XM_011523151.1 | XP_011521453.1 | 76 | 13119 | |||
XM_011523152.1 | XP_011521454.1 | 39 | 6946 | |||
XM_011523155.2 | XP_011521457.1 | 32 | 5947 | |||
XM_017023346.1 | XP_016878835.1 | 76 | 13365 | |||
XM_017023347.1 | XP_016878836.1 | 64 | 11604 | |||
XM_017023348.1 | XP_016878837.1 | 64 | 11344 | |||
GenBank mRNA | AK074472.1 | - | 1 | 12 | ||
AL137259.1 | - | 4 | 589 | |||
BC043273.2 | - | 11 | 1913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
