siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 76, 77 | 13224 | ||
XM_006721206.2 | XP_006721269.1 | 76, 77 | 13118 | |||
XM_011523146.2 | XP_011521448.1 | 77, 78 | 13253 | |||
XM_011523147.1 | XP_011521449.1 | 77, 78 | 13220 | |||
XM_011523148.1 | XP_011521450.1 | 77, 78 | 13293 | |||
XM_011523151.1 | XP_011521453.1 | 76, 77 | 13150 | |||
XM_011523152.1 | XP_011521454.1 | 39, 40 | 6977 | |||
XM_011523155.2 | XP_011521457.1 | 32, 33 | 5978 | |||
XM_017023346.1 | XP_016878835.1 | 76, 77 | 13396 | |||
XM_017023347.1 | XP_016878836.1 | 64, 65 | 11635 | |||
XM_017023348.1 | XP_016878837.1 | 64, 65 | 11375 | |||
GenBank mRNA | AK074472.1 | - | 1, 2 | 43 | ||
AL137259.1 | - | 4, 5 | 620 | |||
BC043273.2 | - | 11, 12 | 1944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
