siRNA Details
Gene Symbol: LOC202181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026921.1 | 6 | 2311 | |||
GenBank mRNA | AK021623.1 | - | 1 | 694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
