siRNA Details
Gene Symbol: C8orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256598.1 | NP_001243527.1 | 5 | 730 | ||
GenBank mRNA | AJ276240.1 | - | 5 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
