siRNA Details
Gene Symbol: GLOD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080489.2 | NP_001073958.2 | 3 | 280 | ||
XM_011543911.2 | XP_011542213.1 | 4 | 383 | |||
XM_011543912.2 | XP_011542214.1 | 4 | 527 | |||
GenBank mRNA | DY655159.1 | - | 2 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
