siRNA Details
Gene Symbol: LRRC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291006.1 | NP_001277935.1 | 8 | 973 | ||
NR_027006.1 | 9 | 2412 | ||||
NR_110991.1 | 7 | 2076 | ||||
XM_011530173.2 | XP_011528475.1 | 8 | 952 | |||
XM_011530175.1 | XP_011528477.1 | 6 | 726 | |||
GenBank mRNA | AK128646.1 | - | 9 | 2391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
