siRNA Details
Gene Symbol: LRRC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291006.1 | NP_001277935.1 | 7, 8 | 952 | ||
XM_011530173.2 | XP_011528475.1 | 7, 8 | 931 | |||
XM_011530175.1 | XP_011528477.1 | 5, 6 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
