siRNA Details
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004522.2 | NP_004513.1 | 26 | 5169 | ||
NR_111932.1 | 20 | 4398 | ||||
XM_011511157.2 | XP_011509459.1 | 19 | 4250 | |||
GenBank mRNA | AB011103.1 | - | 26 | 5138 | ||
AB209191.1 | - | 7 | 3048 | |||
AK126689.1 | - | 1 | 3337 | |||
AL050070.1 | - | 1 | 1543 | |||
AL117393.1 | - | 4 | 2236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
