siRNA Details
Gene Symbol: TPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024458.1 | 10, 11 | 1068 | |||
GenBank mRNA | AF318337.1 | - | 10, 11 | 1068 | ||
AK124332.1 | - | 7, 8 | 1573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
