siRNA Details
Gene Symbol: TPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024458.1 | 11 | 1090 | |||
GenBank mRNA | AF318337.1 | - | 11 | 1090 | ||
AK124332.1 | - | 8 | 1595 | |||
AK127569.1 | - | 3 | 2441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
