siRNA Details
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 4 | 619 | ||
GenBank mRNA | AK097481.1 | - | 4 | 622 | ||
AK302567.1 | - | 4 | 614 | |||
AK302748.1 | - | 4 | 603 | |||
BC070103.1 | - | 4 | 587 | |||
BC094790.1 | - | 4 | 590 | |||
BC117335.1 | - | 4 | 619 | 40125886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
