siRNA Details
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 8, 9 | 1310 | ||
GenBank mRNA | AK097481.1 | - | 8, 9 | 1313 | ||
AK302567.1 | - | 7, 8 | 2528 | |||
AK302748.1 | - | 8, 9 | 1294 | |||
BC070103.1 | - | 8, 9 | 3091 | |||
BC094790.1 | - | 7 | 3257 | |||
BC117335.1 | - | 8, 9 | 1310 | 40125886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
