siRNA Details
Gene Symbol: DKFZP434I0714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033797.2 | 1 | 1535 | |||
GenBank mRNA | AL137273.1 | - | 1 | 1535 | ||
BC034819.1 | - | 1 | 1506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
