siRNA Details
Gene Symbol: DISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020780.1 | NP_065831.1 | 18 | 3526 | ||
XM_011541828.2 | XP_011540130.1 | 17 | 5628 | |||
XM_011541829.1 | XP_011540131.1 | 17 | 2501 | |||
GenBank mRNA | AB037758.1 | - | 18 | 3526 | ||
AB593117.1 | - | 18 | 3609 | |||
AL117235.1 | - | 2 | 165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
