siRNA Details
Gene Symbol: MIR31HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027054.1 | 4 | 605 | |||
GenBank mRNA | AK124391.1 | - | 4 | 606 | ||
BC011715.2 | - | 1 | 120 | |||
BC021861.1 | - | 3 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
