siRNA Details
Gene Symbol: SLC6A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003841.2 | NP_001003841.1 | 12 | 3261 | ||
GenBank mRNA | AK091954.1 | - | 1 | 691 | ||
AK096054.1 | - | 11 | 3269 | |||
AK123779.1 | - | 4 | 1998 | |||
AK290811.1 | - | 12 | 3259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
