siRNA Details
Gene Symbol: SRGAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271872.1 | NP_001258801.1 | 10 | 2763 | ||
XM_005277497.3 | XP_005277554.1 | 10 | 2861 | |||
XM_005277499.4 | XP_005277556.1 | 7 | 1760 | |||
XM_011542015.2 | XP_011540317.1 | 9 | 2334 | |||
XM_017002149.1 | XP_016857638.1 | 8 | 2505 | |||
GenBank mRNA | BC112927.1 | - | 10 | 2763 |
Gene Symbol: SRGAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271870.2 | NP_001258799.2 | 9 | 2624 | ||
XM_005277421.3 | XP_005277478.1 | 7 | 2347 | |||
XM_005277422.4 | XP_005277479.1 | 7 | 1761 | |||
XM_017002085.1 | XP_016857574.1 | 9 | 2700 | |||
XM_017002086.1 | XP_016857575.1 | 9 | 2549 | |||
XM_017002088.1 | XP_016857577.1 | 10 | 2764 | |||
XM_017002089.1 | XP_016857578.1 | 11 | 2812 | |||
XM_017002090.1 | XP_016857579.1 | 10 | 2761 | |||
XM_017002091.1 | XP_016857580.1 | 11 | 2809 | |||
XM_017002092.1 | XP_016857581.1 | 7 | 2344 |
Gene Symbol: SRGAP2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120535.1 | 8 | 2482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
