siRNA Details
Gene Symbol: LINC00917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024406.1 | 4 | 1322 | |||
GenBank mRNA | AK093244.1 | - | 4 | 1322 | ||
BC043250.1 | - | 3 | 1379 | |||
BC144028.1 | - | 2 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
