siRNA Details
Gene Symbol: OTUD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015207.1 | NP_056022.1 | 8 | 1853 | ||
XM_005245792.3 | XP_005245849.1 | 8 | 2512 | |||
XM_005245793.3 | XP_005245850.1 | 7 | 2352 | |||
XM_005245794.3 | XP_005245851.1 | 7 | 1488 | |||
GenBank mRNA | AB007928.3 | - | 8 | 1830 | ||
BC119636.2 | - | 4 | 1071 | |||
BC134416.1 | - | 4 | 1071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
